oxolinic acid
Product Details & Documents
Oxolinic acid is an antibiotic in the quinolone class. It works by inhibiting the bacterial DNA gyrase enzyme, which is essential for DNA replication and repair in bacteria. This disruption of DNA processes ultimately leads to the death of the bacteria.
Caution: Research or further manufacturing use only, not for food or drug use.
- CAS Number:14698-29-4
- Formula:C₁₃H₁₁NO₅
- MDL Number:MFCD00056775
- Molecular Weight:261.233 g/mol
- Storage Temperature:Refrigerator
- EINECS:238-750-8
Specifications
Specification Test Results
- Appearance:White to off white crystalline powder
- Arsenic (As):≤ 20 ppm
- Assay (GC):≥ 99.0 %
- Assay (HPLC):≥ 95.0 %
- Chloride (Cl):≤ 0.012 %
- Heavy Metals:≤ 20 ppm
- Identification (IR):Conforms
- Loss on Drying:≤ 0.3 %
- Melting Point:313 - 317 oC
- Residue on Ignition:≤ 0.2 %
- Solubility (50mg/ml in 0.5M NaOH):Clear, colorless to pale yellow
- BSE/TSE Free:Conforms
Specifications
- Size:100 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



