Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Erlenmeyer Flasks, Closed System

Erlenmeyer flask with Bi-directional transfer cap, TPE tube, heat seal, 250 ml, Sterile, ¹/₈", ¹/₄", 50 cm
  77509-104
 :  C30921-AZC050A
Erlenmeyer Flasks, Closed System
  77509-104
Erlenmeyer Flasks, Closed System

Erlenmeyer flask with Bi-directional transfer cap, TPE tube, heat seal, 250 ml, Sterile, ¹/₈", ¹/₄", 50 cm

 
$486.40
   
Case of 4
 

For aseptic transfer of culture medium and cell culture solution.

  • Filter membrane area (0.22 μm): Ø 42 mm
  • Tube material: TPE

One end of the pipeline is connected to the MPC series connector, which increases the convenience and safety in use. The male connector and the sealing cap are tightly connected to prevent accidental disconnection, and the connector can be rotated to reduce pipeline distortion.

 
  • Sterility:
    Sterile
  • Capacity:
    250 ml
  • Description:
    Erlenmeyer flask with Bi-directional transfer cap, TPE tube, heat seal
  • Neck I.D.:
    ¹/₈"
  • Neck O.D.:
    ¹/₄"
  • Tubing length:
    50 cm