Orders LinkContactUsLinkComponent
Inoculating Needle/Loops
Inoculating Needle/Loops
Inoculating loop, 1 μl, Blue, 400/pack, 4000/case
Catalog # 77508-608
Supplier:   NEST SCIENTIFIC
Supplier #:  717101
Inoculating Needle/Loops
Catalog # 77508-608
Supplier:  NEST SCIENTIFIC
Inoculating Needle/Loops
Inoculating loop, 1 μl, Blue, 400/pack, 4000/case
Price
$488.55
Log in for availability
Case of 4,000
Product Details & Documents
Inoculating needle/loops made up of PP material, hydrophilic.
  • Applied in microbiology experiments
  • Different colors for identification
  • Sterilized by ethylene oxide
  • Easy-tear sterile individual packaging
Flexible handles for easier collection and inoculation.
Specifications
  • Color:
    Blue
  • Description:
    Inoculating loop, 1 μl
  • Packaging:
    400/pack, 4000/case