Orders LinkContactUsLinkComponent
Colored Cap Inserts
Colored Cap Inserts
Coloured cap insert, Assorted
  77508-338
Supplier #:  611007
Colored Cap Inserts
  77508-338
Colored Cap Inserts
Coloured cap insert, Assorted
$40.18
Case of 600
Six colored cap inserts for easy identification. Compatible with regular cryogenic vials that with external thread caps only.
Packaging: 100/pack; 1000/case
  • Color:
    Assorted
  • Description:
    Coloured cap insert