Orders LinkContactUsLinkComponent
Colored Cap Inserts
Colored Cap Inserts
Coloured cap insert, Yellow
Catalog # 77508-330
Supplier:   NEST SCIENTIFIC
Supplier #:  611003
Colored Cap Inserts
Catalog # 77508-330
Supplier:  NEST SCIENTIFIC
Colored Cap Inserts
Coloured cap insert, Yellow
Price
$62.77
Log in for availability
Case of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Six colored cap inserts for easy identification. Compatible with regular cryogenic vials that with external thread caps only.
Packaging: 100/pack; 1000/case
Specifications
  • Color:
    Yellow
  • Description:
    Coloured cap insert