Colored Cap Inserts
Coloured cap insert, Red
77508-328
Supplier #: 611002
Six colored cap inserts for easy identification. Compatible with regular cryogenic vials that with external thread caps only.
Packaging: 100/pack; 1000/case
- Color:Red
- Description:Coloured cap insert