Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Pasteur Pipettes

Pasteur Pipettes

Pasteur pipette, PP, 2 ml, 150 mm, Sterile, 2000/case
Catalog # 77507-584
Supplier:   NEST SCIENTIFIC
Supplier #:  318112
Pasteur Pipettes
Catalog # 77507-584
Supplier:  NEST SCIENTIFIC
Pasteur Pipettes

Pasteur pipette, PP, 2 ml, 150 mm, Sterile, 2000/case

Price
$226.61
Log in for availability
Case of 2,000
Product Details & Documents

Pipette can be twisted for use with micro vessels and special-shaped containers.

  • Good chemical stability
  • Transparent with clear graduations for easy observation
  • High precision and good repeatability
  • Individually wrapped, sterilized by EO
  • DNase/RNase, endotoxin and pyrogenic free
Specifications
  • Sterility:
    Sterile
  • Volume:
    2 ml
  • Description:
    Pasteur pipette
  • Length:
    150 mm
  • Material:
    PP
  • Packed:
    2000/case
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.