Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Pipette Tips

Graduated, 5 ml, +, Clear, For Thermo, 400/pack, 720/case
Catalog # 77507-528
Supplier:   NEST SCIENTIFIC
 :  307022
Pipette Tips
Catalog # 77507-528
Supplier:  NEST SCIENTIFIC
Pipette Tips

Graduated, 5 ml, +, Clear, For Thermo, 400/pack, 720/case

Price
$413.59
Log in for availability
Case of 1,200
Product Details & Documents

NEST 5 and 10 ml pipette tips are precision-molded from high purity, DNase RNase and pyrogen free, virgin polypropylene.

  • Autoclavable at 121 ℃
  • Meet a wider range of volumetric pipetting needs for more precise pipetting

They are available in 5 and 10 ml, filtered or non-filtered. Choose from compatible pipettors either for Eppendorf, Sartorius, Gilson, or for Thermo pipettors.

They allow pipetting from narrow volume measuring equipment. Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.

 : ISO Class 08.

Specifications
  • Volume:
    5 ml
  • Tip color:
    Clear
  • Packed:
    400/pack, 720/case
  • Sterile:
    +
  • Tip style:
    Graduated
  • For use with:
    For Thermo