FluidX 1.6 ml Tri-coded Tubes, 48-format, External Thread
Product Details & Documents
FluidX 1.6 ml maximum recovery tube is ideally suited for automated workflows using liquid handling, where more sample can be retrieved due to the unique shaped bottom.
- Sample identification from 1D, 2D and human readable code
- Capability to work with automation, designed for liquid handlers
- Suitable for cryogenic storage
- Made from high-quality virgin PP
- Available in 48-well format SBS racks or bulk
- 480 tubes per case or 10 racks per case
Tri-coded tubes offer unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities.
Suitable for use with liquid handling automation, whether reformatting, cherry picking, aliquoting an/or normalization. The 1.6 ml maximum recovery tube offers unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities.
Designed and developed with broad compatibility in mind, the tubes perform without compromise in conjunction with automated barcode reading, capping and sample management systems from Azenta Life Sciences and all other industry-recognized manufacturers.
Storage temperature: Minimum temperature (with screw cap): −196 °C (gas phase liquid nitrogen)
Applications: (For research use only) Liquid handling workflows, biobanking, cryogenic storage, cancer research, cellular and molecular biology, drug discovery, compound libraries, reagent storage, CDMOs.
Documents
FluidX 1.6 ml Tri-coded Tubes, 48-format, External Thread
Application Notes
Specifications
- Cap included:No
- Capacity:1.6 ml
- Packaging:Bulk
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



