Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GLOVE EXAM LATEX CURAD PF MD 21LB CS1000
Catalog # MSPP-4035
Supplier:
ASSAY CELL TECHNOLOGIES INC MS
Expand image
GLOVE EXAM LATEX CURAD PF MD 21LB CS1000
Catalog # MSPP-4035
Supplier:
ASSAY CELL TECHNOLOGIES INC MS
Supplier Number:
4035