Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TAG EYEWASH STATION INSPECTN VINYL PK25
Catalog # MSPP-S-21108
Supplier:
ULINE MS
Expand image
TAG EYEWASH STATION INSPECTN VINYL PK25
Catalog # MSPP-S-21108
Supplier:
ULINE MS
Supplier Number:
S-21108