Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
BIOXP® Select mRNA

BIOXP® Select mRNA

Kit for BioXp 3250, psuedouridine modifications, 24
Catalog # 77461-276
Supplier:   TELESIS BIO
Supplier #:  BX-MRNA-RSM1-CC-24
BIOXP® Select mRNA
Catalog # 77461-276
Supplier:  TELESIS BIO
CAS Number:  
BIOXP® Select mRNA
BIOXP® Select mRNA

Kit for BioXp 3250, psuedouridine modifications, 24

CAS Number:  
Price
$4,515.91
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The BioXp Select mRNA synthesis kit enables the synthesis of mRNA from an input DNA template.

  • mRNA Length: 0.4 to 10 kb
  • Yield: >50 µg/well
  • Compatible with BioXp 3250
  • Cap: CleanCap® AG (3'OMe)
  • Modification: Pseudouridine

Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.

Specifications

Specifications

  • Description:
    Kit for BioXp 3250, psuedouridine modifications
  • No. of reactions:
    24

Specifications