Orders LinkContactUsLinkComponent
BioXp® Tip Kits
BioXp® Tip Kits
5×50 µl rack, 3×200 µl rack
  77461-214
Supplier #:  12214KT
BioXp® Tip Kits
  77461-214
BioXp® Tip Kits
BioXp® Tip Kits
5×50 µl rack, 3×200 µl rack
$122.57
Pack of 1
Minimum order quantity is 1
BioXp® system tips in various kit types and sizes.
  • Free of human DNA
  • Rnase/Dnase free
  • PCR inhibitor free
  • Automated
Different kit types and sizes will require different quantities of 50 and 200 µl racks.
Ordering information: Verify the tip requirements for the BioXp® kit that you are ordering.
  • Description:
    5×50 µl rack, 3×200 µl rack