Orders LinkContactUsLinkComponent
BioXp® Tip Kits
8×50 µl rack, 1×200 µl rack
  77461-208
 :  12211KT
BioXp® Tip Kits
  77461-208
BioXp® Tip Kits
BioXp® Tip Kits
8×50 µl rack, 1×200 µl rack
$137.16
Pack of 1
BioXp® system tips in various kit types and sizes.
  • Free of human DNA
  • Rnase/Dnase free
  • PCR inhibitor free
  • Automated
Different kit types and sizes will require different quantities of 50 and 200 µl racks.
 : Verify the tip requirements for the BioXp® kit that you are ordering.
  • Description:
    8×50 µl rack, 1×200 µl rack