Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
BIOXP® Select Plasmid AMP

BIOXP® Select Plasmid AMP

Kit for BioXp 3250, 8
Catalog # 77461-294
Supplier:   TELESIS BIO
Supplier #:  BX-SEL-PLSAMP-08
BIOXP® Select Plasmid AMP
Catalog # 77461-294
Supplier:  TELESIS BIO
CAS Number:  
BIOXP® Select Plasmid AMP
BIOXP® Select Plasmid AMP

Kit for BioXp 3250, 8

CAS Number:  
Price
$1,254.42
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

BioXp Select plasmid amplification kits contain reagents for isothermal amplification of plasmid DNA.

  • Input: Plasmids ranging from 1.8 to 37 kb
  • Yield: >10 µg/well
  • Output format: Branched concatemers
  • Compatible with BioXp 3250

Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.

Specifications

Specifications

  • Description:
    Kit for BioXp 3250
  • No. of reactions:
    8

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.