Orders LinkContactUsLinkComponent
BioXp® Tip Kits
BioXp® Tip Kits
8×50 µl rack, 2×200 µl rack
Catalog # 77461-192
Supplier:   TELESIS BIO
Supplier #:  12203KT
BioXp® Tip Kits
Catalog # 77461-192
Supplier:  TELESIS BIO
BioXp® Tip Kits
BioXp® Tip Kits
8×50 µl rack, 2×200 µl rack
Price
$151.76
Log in for availability
Pack of 1
Minimum order quantity is 1
Product Details & Documents
BioXp® system tips in various kit types and sizes.
  • Free of human DNA
  • Rnase/Dnase free
  • PCR inhibitor free
  • Automated
Different kit types and sizes will require different quantities of 50 and 200 µl racks.
Ordering information: Verify the tip requirements for the BioXp® kit that you are ordering.
Specifications
  • Description:
    8×50 µl rack, 2×200 µl rack