Orders LinkContactUsLinkComponent
NEXTFLEX™ UDI Barcodes
NEXTFLEX™ UDI barcodes (1 - 96), 192
  77448-162
 :  NOVA-514150
NEXTFLEX™ UDI Barcodes
  77448-162
 :  
NEXTFLEX™ UDI Barcodes
NEXTFLEX™ UDI Barcodes
NEXTFLEX™ UDI barcodes (1 - 96), 192
 :  
$2,984.94
Each
The NEXTFLEX® Unique dual index barcodes are barcoded adapters that can be used with single or paired-end read sequencing library prep kits  designed for sequencing on Illumina® platforms workflows.
  • 384 UDI adapters for mitigating sample misassignment due to index hopping
  • Unique dual index adapters for multiplexing up to 384 libraries for Illumina® sequencing platforms
  • Mitigate index hopping or spread of signal that can occur during sequencing
  • Each lot of the NEXTFLEX® Unique dual index barcodes is tested for index purity by sequencing
  • Compatible with both paired-end and single-read sequencing
  • Compatible with the NEXTFLEX® Universal blockers
The NEXTFLEX® Unique dual index barcodes utilize two unique 8 nt indexes on each UDI adapter. The 8 nt indices are designed with a Hamming Distance of at least three throughout the entire series, permitting error correction. Patterned flow cells, such as those found on the Illumina® NovaSeq® and HiSeq® 3000/4000 platforms have been recognized to suffer from increased rates of sample mis-assignment during sequencing.
The use of adapters with unique dual indices during sequencing prevents such mis-assigned reads from appearing in final data sets, allowing for the highest assurance of data integrity. The NEXTFLEX® Unique dual index barcodes are barcoded adapters, for sequencing on Illumina® platforms, that provide data security in sequencing applications. Multiplexing with NEXTFLEX® Unique dual index barcodes increase scale while reducing costs by allowing the user to pool up to 384 libraries in a single flow cell lane.
 : Manufactured in ISO 9001 facility.
 : For research use only. Not for use in diagnostic procedures.
  • Description:
    NEXTFLEX™ UDI barcodes (1 - 96)
  • No. of reactions:
    192