NEXTFLEX™ UDI Barcodes
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The NEXTFLEX® Unique dual index barcodes are barcoded adapters that can be used with single or paired-end read sequencing library prep kits designed for sequencing on Illumina® platforms workflows.
- 384 UDI adapters for mitigating sample misassignment due to index hopping
- Unique dual index adapters for multiplexing up to 384 libraries for Illumina® sequencing platforms
- Mitigate index hopping or spread of signal that can occur during sequencing
- Each lot of the NEXTFLEX® Unique dual index barcodes is tested for index purity by sequencing
- Compatible with both paired-end and single-read sequencing
- Compatible with the NEXTFLEX® Universal blockers
The NEXTFLEX® Unique dual index barcodes utilize two unique 8 nt indexes on each UDI adapter. The 8 nt indices are designed with a Hamming Distance of at least three throughout the entire series, permitting error correction. Patterned flow cells, such as those found on the Illumina® NovaSeq® and HiSeq® 3000/4000 platforms have been recognized to suffer from increased rates of sample mis-assignment during sequencing.
The use of adapters with unique dual indices during sequencing prevents such mis-assigned reads from appearing in final data sets, allowing for the highest assurance of data integrity. The NEXTFLEX® Unique dual index barcodes are barcoded adapters, for sequencing on Illumina® platforms, that provide data security in sequencing applications. Multiplexing with NEXTFLEX® Unique dual index barcodes increase scale while reducing costs by allowing the user to pool up to 384 libraries in a single flow cell lane.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Documents
NEXTFLEX™ UDI Barcodes
Application Notes
Specifications
Specifications
- Description:NEXTFLEX™ UDI barcodes (193 - 288)
- No. of reactions:192



