Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
NEXTFLEX™ 16S V1-V3 Amplicon-Sequence Kits

NEXTFLEX™ 16S V1-V3 Amplicon-Sequence Kits

NEXTFLEX™ 16S V1-V3 (1 - 48), 96
Catalog # 77448-090
Supplier #:  NOVA-4202-03
NEXTFLEX™ 16S V1-V3 Amplicon-Sequence Kits
Catalog # 77448-090
CAS Number:  
NEXTFLEX™ 16S V1-V3 Amplicon-Sequence Kits
NEXTFLEX™ 16S V1-V3 Amplicon-Sequence Kits

NEXTFLEX™ 16S V1-V3 (1 - 48), 96

CAS Number:  
Price
$2,220.58
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The NEXTFLEX® 16S V1-V3 Amplicon-Sequence kit is designed to prepare multiplexed amplicon libraries that span the hypervariable domains one to three (V1-V3) of microbial 16S ribosomal RNA (rRNA) genes.

  • Fast and easy library prep protocol
  • Low input - Only 1 ng of input required
  • Multiplex up to 384 libraries
  • Custom sequencing primers are not required
  • Automation protocols are now available for the Sciclone® NGS and NGSx Workstations to automate your 16S sequencing
  • Functionally tested with Illumina® sequencing platforms

Optimized protocol offers lower PCR bias and fewer off-target reads: The NEXTFLEX® 16S V1-V3 Amplicon-Seq kit incorporates a second PCR step in the protocol for the addition of the sample-specific index which reduces off-target reads encountered during amplicon sequencing.

384 Barcodes available for cost-effective 16S V1-V3 rRNA sequencing: The NEXTFLEX 16S V1-V3 Amplicon-Seq library prep kit can be used for multiplexing up to 384 samples, to greatly reduce the cost of sequencing. The barcodes are offered in sets of 12, 48 or 96 barcodes and supplied with 2 reactions worth of each barcode.

Automate your 16S V1-V3 Library Prep to Increase Throughput and Reduce Errors
A NEXTFLEX® automation protocol is now available on the Sciclone® NGS and NGSx workstations to help labs increase their throughput and reduce human errors.

The NEXTFLEX 16S V1-V3 rRNA Amplicon-seq panels are now available bundled with access to CosmosID-HUB; an online software solution that enables fast and easy analysis of complex microbiome data. CosmosID-HUB gives scientists user-friendly access to version-controlled and validated 16S pipelines. The machine-learning powered software also enables rapid data interpretation through a comparative analysis software with features such as tables, heatmaps, bar charts, multiple Alpha & Beta Diversity indexes, abundance distribution plots, differential abundance testing, as well as comprehensive statistics between groups. Learn more about CosmosID-HUB.

Certifications: Manufactured in ISO 9001 facility.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    NEXTFLEX™ 16S V1-V3 (1 - 48)
  • No. of reactions:
    96

Specifications