$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
BUFFER KIT PROTEIN REACTION CLICK-GO
MSPP-CCT-1262
:
Vector Laboratories
BUFFER KIT PROTEIN REACTION CLICK-GO
MSPP-CCT-1262
:
Vector Laboratories
:
CCT-1262