Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RACK COMORACK-96 W/COVER PC PK10
Catalog # MSPP-MP22504
Supplier:
NBS SCIENTIFIC, LLC MS
Expand image
RACK COMORACK-96 W/COVER PC PK10
Catalog # MSPP-MP22504
Supplier:
NBS SCIENTIFIC, LLC MS
Supplier Number:
MP22504