$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
TUBE N-CODED SCREW CAP 1.10ML PK960
MSPP-MP32033L
:
NBS SCIENTIFIC, LLC MS
TUBE N-CODED SCREW CAP 1.10ML PK960
MSPP-MP32033L
:
NBS SCIENTIFIC, LLC MS
:
MP32033L