Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RACK 96-4 W/LOW COVER F/1.40ML TUBE PK10
Catalog # MSPP-MP51210
Supplier:
NBS SCIENTIFIC, LLC MS
Expand image
RACK 96-4 W/LOW COVER F/1.40ML TUBE PK10
Catalog # MSPP-MP51210
Supplier:
NBS SCIENTIFIC, LLC MS
Supplier Number:
MP51210