Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
BUFFER BLOCKING V20 500ML

BUFFER BLOCKING V20 500ML

Catalog # 77442-244
Supplier:   Teknova
Supplier #:  2B2005
BUFFER BLOCKING V20 500ML
Catalog # 77442-244
Supplier:  Teknova
CAS Number:  
BUFFER BLOCKING V20 500ML
CAS Number:  
Price
$101.52
Log in for availability
Each
Minimum order quantity is 8
Product Details & Documents