Orders LinkContactUsLinkComponent
CTAB Extraction Solution
CTAB extraction solution, sterile, 250 ml
  77443-472
 :   Teknova
 :  C2190
CTAB Extraction Solution
  77443-472
 :  Teknova
 :  
CTAB Extraction Solution
CTAB extraction solution, sterile, 250 ml
 :  
$101.52
Each
CTAB is a cationic detergent used in the extraction of genomic DNA from bacteria and for the isolation of high molecular weight DNA from plants for plamid minipreps and for PCR.
The Solution contains 1% CTAB, 100 mM Tris-HCl (pH 8,0), 20 mM EDTA (pH 8,0) and 1.4 M Sodium Chloride.
  • Description:
    CTAB extraction solution, sterile
  • Size:
    250 ml