Explorer™ Precision Balances
EX6201N/E, 6200 g, 190×200 mm (7.5×8"), 0.1 g
Catalog # 77435-014
Supplier: Ohaus
: 30553891
Jump to:
Product Details & Documents
The Explorer series is the flagship precision balance range from OHAUS. Equipped with a weighing cell that's precision-machined from a solid metal block, Explorer balances are extremely accurate and durable, and are ideal for laboratory and industrial environments.
- Four-level user management with password protection, and un-editable system log plus a high level of configurability make Explorer balances ideal for regulated applications
- Detachable terminal with a large color touchscreen and programmable IR sensors
- Frameless draftshield make Explorer precision balances easy and convenient to use
The OHAUS Explorer Series of precision balances combine modern features and design elements to offer unmatched functionality in a line of high-performance balances unlike any other on the market. These smart and intuitive balances simplify even the most complex laboratory measurements.
Specifications
- Model:EX6201N/E
- Pan/Platform Size:190×200 mm (7.5×8")
- Readability:0.1 g
- Weighing capacity:6200 g



