2'-Methylacetophenone
Bottle, 100 g
77403-868
: OR11271-100G
: o-Methylacetophenone, Methyl o-tolyl ketone, 1-(o-Tolyl)ethanone
- :577-16-2
- :C₉H₁₀O
- :MFCD00008734
- :134.18 g/mol
- :214 °C
- :1.026 g/cm³ (25 °C)
- :75 °C
- :Ambient
- :209-408-5
- Pack type:Bottle
- Size:100 g