BRAND® Transferpette® -8/-12 Multi-Channel Electronic Pipettes, Trade-Up Promotion
Catalog # MP77322-326
Supplier: Brandtech
Specifications
- Volume:50 - 1250 µl
- For use with:50 - 1250 µl tips
- Increments:1.0 µl
- Accuracy:±1%
- Precision:≤0.25%
Specifications
About this item
Transferpette® electronic pipettes are lightweight, ergonomic, fully autoclavable, and work with a wide range of tip brands.
True one-hand operation for right- and left-handed users; easily set the volume, pipette, and eject the tip
- Color code on volume adjustment wheel, shaft, and volume-change protection
- Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
- Easy insertion and ejection of tips due to O-rings on multi-channel pipettes
- Universal tip cone
- Fully autoclavable at 121 °C
The volume-change protection is easy to operate with gloves on, and protects the set volume against accidental adjustment. The Transferpette® electronic also features easy calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that the correct pipette tips can always be ordered.
Delivery:
Supplied with performance certificate, battery, power adapter, reagent reservoir, installation tool, and silicone grease.