Contec® Compact Bucket Systems
Product Details & Documents
The autoclavable Compact Bucket System is an all-in-one solution to make cleaning and disinfecting easier and more efficient. The cart is designed for use with Contec® mops such as the VertiKlean® MAX™, VertiKlean® MAX™ Sealed Edge, EasyCurve®, and QuickTask™ mopping systems.
On-cart storage for two mop handles and frames allows convenient storage and keeps mops out of the way
- Mop holders keep mops securely in place when changing locations
- Improved castors and connections significantly reduces noise during use, allowing more durable attachments with less 'rattle'
- Compact polypropylene buckets with integral drip cap stay in place while in use, preventing drips between the buckets and floor
- Buckets can hold a curved or flat wringer for curved or flat mops
- Integral mop head removal tool or optional hands-free mop head removal tool allows easy removal of mop heads
- 100% autoclavable, electropolished stainless steel cart is safe for autoclave at 121 °C for 30 minutes
- Cart can be used in the most critical environments, and is compatible with most cleanroom disinfectants and cleaners
- Utility handle basket holds 2×6 liter buckets; providing additional storage for clean mop heads, solution bottles, wipes, and other cleaning supplies
The stainless steel cart is available in multiple configurations to meet the unique needs of any environment. It is available with either two or three autoclavable buckets and a utility cart handle. Each cart is fitted with quiet, easy to move casters that lock in place when needed. The utility handle allows two small buckets (1.6 gallon) or one large bucket (3.9 gallon) to be positioned on the handle, providing additional storage for extra supplies such as clean mop heads, solution spray bottles, wipes, and other items as needed.
The cart frame features an integrated mop head removal tool on the front for easy, hands-free removal of a VertiKlean® MAX™ or VertiKlean® MAX™ Sealed Edge mop head. An optional hands-free removal tool is also available for both the VertiKlean® MAX mop heads and the QuickTask™ mop heads. Two mop holders with mop brace and handle clasps on either side of the cart keep the mop in position and allow for convenient mop storage during transport.
The systems are provided with stainless steel debris screens. The screen is positioned in the base of the clean and rinse buckets and prevents the mop from touching the bottom of the bucket where unwanted contaminants settle.
Specifications
- Capacity:2×15 L (3.9 gal.), 2×6 L (1.6 gal.)
- Description:Compact bucket system with PP buckets (two 15 L, two 6 L), utility handle, and one debris screen
Accessories
Product Information
- Catalog #ColorCapacityAvailabilityPrice
- Red, blue and gray polypropylene autoclavable bucketsRed, blue and gray polypropylene autoclavable buckets76199-896Assorted15 L (3.9 gal.)Log in for availabilityCase of 3 - $170.00
Product Information
- Catalog #MaterialFor Use WithAvailabilityPrice
- Flat Mop WringerFlat Mop Wringer76199-932Stainless steelVertiKlean® MAX™ and QuickTask™ Mops (15 l (3.9 gal.) and 25 l (6.5 gal.) Buckets)Log in for availabilityCase of 1 - $420.00
Product Information
- Catalog #ColorCapacityAvailabilityPrice
- Polypropylene 6L utility bucketPolypropylene 6L utility bucket76199-890Blue6 L (1.6 gal.)Log in for availabilityCase of 2 - $75.80
Product Information
- Catalog #AvailabilityPrice
- VertiKlean® MAX hands-free mop removal tool for compact bucket systemVertiKlean® MAX hands-free mop removal tool for compact bucket system76311-188Log in for availabilityCase of 1 - $344.00
Product Information
- Catalog #AvailabilityPrice
- QuickTask™ hands-free removal tool and bag holder
Product Information
- Catalog #DimensionsAvailabilityPrice
- Cleanroom disposal bag
Product Information
- Catalog #AvailabilityPrice
- Stainless Steel Connecting Kit for Multi-Bucket SystemStainless Steel Connecting Kit for Multi-Bucket System89073-986Log in for availabilityEach - $97.10
Product Information
- Catalog #AvailabilityPrice
- Stainless steel debris screen for 15L polypropylene bucketsStainless steel debris screen for 15L polypropylene buckets76199-892Log in for availabilityCase of 2 - $175.00
Product Information
- Catalog #MaterialFor Use WithAvailabilityPrice
- Curved Mop WringerCurved Mop Wringer76199-888Stainless steelEasyCurve® Mops (15 l (3.9 gal.) and 25 l (6.5 gal.) Buckets)Log in for availabilityCase of 1 - $468.00
Product Information
- Catalog #CapacityAvailabilityPrice
- Bucket bag liners for polypropylene buckets, irradiatedBucket bag liners for polypropylene buckets, irradiated76199-89415 L (3.9 gal.)Log in for availabilityCase of 60 - $418.00