Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CLONE G-PROTEIN ALPHA INHIBITR-1 HU 10UG
Catalog # MSPP-SC309030
Supplier:
Origene Technologies
Expand image
CLONE G-PROTEIN ALPHA INHIBITR-1 HU 10UG
Catalog # MSPP-SC309030
Supplier:
Origene Technologies
Supplier Number:
SC309030