$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
TUBE SCREW CAP MATRIX CODED 1.40ML PK10
MSPP-MP52761SY
:
NBS SCIENTIFIC, LLC MS
TUBE SCREW CAP MATRIX CODED 1.40ML PK10
MSPP-MP52761SY
:
NBS SCIENTIFIC, LLC MS
:
MP52761S-Y20