Orders LinkContactUsLinkComponent
511 Basic Spectacles
511 Basic Spectacles
Anti-fog, scratch-resistant, Clear, Black
  77393-754
Supplier #:  MDU5033S
511 Basic Spectacles
  77393-754
511 Basic Spectacles
511 Basic Spectacles
Anti-fog, scratch-resistant, Clear, Black
$80.42
Pack of 10
The 511 Basic spectacle is designed with a frame made from non-allergic material.
  • Anti-fog and anti-scratch lens treatment
  • Adjustable temples
  • Integrated side shields
Temples are adjustable in length and inclination.
  • Frame color:
    Black
  • Lens coating:
    Anti-fog, scratch-resistant
  • Lens tint:
    Clear