Orders LinkContactUsLinkComponent
546 Value Spectacles
546 Value Spectacles
Anti-fog, scratch-resistant, Clear, Black/orange
Catalog # 77393-744
Supplier #:  MDU5028SC
546 Value Spectacles
Catalog # 77393-744
546 Value Spectacles
546 Value Spectacles
Anti-fog, scratch-resistant, Clear, Black/orange
Price
$125.34
Log in for availability
Pack of 10
Product Details & Documents
Extremely lightweight and modern spectacles with a sporty line.
  • Multiple lens treatments are available, including anti-fog, anti-scratch, and UV525
  • Wraparound polycarbonate lens
  • Integrated side shields
546 Value spectacles feature temples equipped with rubber terminals to guarantee excellent comfort, as well as a non-slip rubber nose pad. The UV525 protects eyes during curing light usage. Smoke lenses can help to reduce anxiety.
Specifications
  • Frame color:
    Black/orange
  • Lens coating:
    Anti-fog, scratch-resistant
  • Lens tint:
    Clear