Orders LinkContactUsLinkComponent
Face Shields, 711
Face Shields, 711
711 face shield, Clear, Clear
Catalog # 77393-774
Supplier #:  MDU7006S
Face Shields, 711
Catalog # 77393-774
Face Shields, 711
Face Shields, 711
711 face shield, Clear, Clear
Price
$26.22
Log in for availability
Each
Product Details & Documents
The 711 series face shields feature an anti-fog lens treatment.
  • Anti-fog lens treatment
  • Disposable and interchangeable shields
  • Ultra-lightweight design
  • Multiple frame colors available
The 711 series face shields can be worn with prescription glasses and protective masks.
Ordering information: Anti-fog replacement shields are available separately.
Specifications
  • Frame color:
    Clear
  • Description:
    711 face shield
  • Lens tint:
    Clear