Spin Dial
Spin dial, 5¼", 3", Stainless steel
Catalog # 77391-940
Supplier: Bio-Serv
Supplier #: K3140
Jump to:
Product Details & Documents
Spin dial is multi-species interactive enrichment device. Smear with honey or peanut butter plus foraging bits for added engagement.
- Made of stainless steel
- Hangs onto cage with a quick link
- Extremely durable
- Autoclavable
- Cagewasher safe
Specifications
- Ø:3"
- Description:Spin dial
- Length:5¼"
- Material:Stainless steel