QuickConnect™ Mop Handles
Product Details & Documents
Interchangeable mop handles are available in carbon fiber, anodized aluminum, stainless steel or fiberglass.
- Compatible with all QuickConnect™ frames
- Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, anodized aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths. For hard-to-reach ceilings and walls, a carbon fiber handle is available that is lightweight and easy to maneuver. A one-piece stainless-steel handle is also available.
Documents
Certificates
Search by Lot NumberAvantor Contec® Products for Mopping Cleanroom Environments.pdf Contec Mopping Products for Critical Environments Brochure v2 Contec Products for Cleaning Small Environments.pdf Contec Products for Research Education and Labs.pdf Production Insight Solutions for Controlled Environments and Manufacturing Processes
Product Literature
Specifications
- Description:QuickConnect™ carbon fiber telescopic handle
- Length:175 - 848 cm (69 - 334")
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement locking collar set for 77322-290
Product Information
- Catalog #AvailabilityPrice
- Replacement locking collar set for 77322-292, 77322-296 and 77322-288Replacement locking collar set for 77322-292, 77322-296 and 77322-28878615-924Log in for availabilityEach - $12.22
Product Information
- Catalog #AvailabilityPrice
- Replacement locking collar set for 77322-294