$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
BUFFER SGVB++ LOW IONIC STRENGTH 250ML
MSPP-B1-10
:
COMPTECH MS
BUFFER SGVB++ LOW IONIC STRENGTH 250ML
MSPP-B1-10
:
COMPTECH MS
:
B110