Orders LinkContactUsLinkComponent
Code Corporation Barcode Reader Accessories
Code Corporation Barcode Reader Accessories
Cable, black, 8' cable length, Code readers
Catalog # 77282-168
Supplier:   Brady Worldwide
Supplier #:  176700
Code Corporation Barcode Reader Accessories
Catalog # 77282-168
Supplier:  Brady Worldwide
Code Corporation Barcode Reader Accessories
Code Corporation Barcode Reader Accessories
Cable, black, 8' cable length, Code readers
Price
$36.80
Log in for availability
Each
Product Details & Documents
Accessories available for the barcode readers.
Use the 6′ (176701) or 8' (176700) Coiled RS232 Affinity® Cable CRA-C511 with Code's cabled barcode readers.
Model 176702 Code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3.3 A).
Model 176703 Code power supply is an international power supply with USB connection.
Model 176856 - Use this stand to scan hands-free or simply to hold your scanner. It features a flexible gooseneck design that adjusts easily to different sized scanners.
Specifications
  • Description:
    Cable, black, 8' cable length
  • For model:
    Code readers