Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
GLOVES NITR HYH PUREZERO GREEN LG CS1000
MP76554-016
:
O&M HALYARD CANADA, INC CA
GLOVES NITR HYH PUREZERO GREEN LG CS1000
MP76554-016
:
O&M HALYARD CANADA, INC CA
:
LFS321LG