$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
GLOVES NIT HYH PUREZERO BLUE SM CS500
MP76554-014
:
O&M HALYARD CANADA, INC CA
GLOVES NIT HYH PUREZERO BLUE SM CS500
MP76554-014
:
O&M HALYARD CANADA, INC CA
:
LFS121SM