Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BP FLEX 2 PH GAS CALIBRATOR CARTRIDGE MV
Catalog # MSPP-58102
Supplier:
Nova Biomedical
Expand image
BP FLEX 2 PH GAS CALIBRATOR CARTRIDGE MV
Catalog # MSPP-58102
Supplier:
Nova Biomedical
Supplier Number:
58102