$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
BP FLEX 2 PH GAS MICROSENSOR CARD HV
MSPP-57526
:
Nova Biomedical
BP FLEX 2 PH GAS MICROSENSOR CARD HV
MSPP-57526
:
Nova Biomedical
:
57526