Orders LinkContactUsLinkComponent
Lighthouse Launderable Microfiber Mops
Lighthouse EIP Hood Cleaning Tool (HCT), adjustable length handle (29" – 53")
  77007-792
 :  THCT521122
Lighthouse Launderable Microfiber Mops
  77007-792
Lighthouse Launderable Microfiber Mops
Lighthouse Launderable Microfiber Mops
Lighthouse EIP Hood Cleaning Tool (HCT), adjustable length handle (29" – 53")
$39.56
Each
The launderable microfiber mop can be used both as a dry and wet mop to clean floors in any facility or environment.
  • Effective in removing dirt and bacteria
  • Saves money and resources
  • Eco-friendly
  • Can be washed and reused
Microfiber mops can remove dirt and grime from floors more effectively than traditional mops because the fibers attract and hold onto dirt particles. Washable microfiber mops can be used for hundreds of cleanings before needing to be replaced, which saves money on purchasing new mops and reduces waste.
 : Mop frame and adjustable handle sold separately.
  • Description:
    Lighthouse EIP Hood Cleaning Tool (HCT), adjustable length handle (29" – 53")