Orders LinkContactUsLinkComponent
Bee-Write™ ESD Cleanroom Papers
Bee-Write™ ESD Cleanroom Papers
ESD cleanroom paper, Blue, 8.5×11", 250 sheets/ream
Catalog # 77008-002
Supplier #:  BW-CRP-8511-BLUE
Bee-Write™ ESD Cleanroom Papers
Catalog # 77008-002
Bee-Write™ ESD Cleanroom Papers
ESD cleanroom paper, Blue, 8.5×11", 250 sheets/ream
Price
$205.89
Log in for availability
Case of 2,500
Product Details & Documents
Bee-Write™ ESD Cleanroom Paper is cleanroom packaged (vacuum packed, double-bagged) and processed in a class 100 ISO 5 controlled environment.
  • Low Chloride and other ionic contamination; ultra-low ionic contamination
  • Formulated without Magnesium Silicate
  • Excellent IPA resistance
  • Compatible to most inks in writing without smearing
  • Ultra-low particle and fiber generation
  • Excellent heat resistance material for laser printing and photocopiers
  • Compatible with laser and inkjet printers
  • Latex-free
  • ROHS compliant
Because Bee-Write™ Cleanroom Paper is ESD safe, it is also suitable for use in a variety of electronics assembly cleanrooms. It is compatible with most pens as well as laser and inkjet printers.
Specifications
  • Color:
    Blue
  • Description:
    ESD cleanroom paper
  • Dimensions:
    8.5×11"
  • Packaging:
    250 sheets/ream