Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Vanillin 99%

Glass bottle, 100 g
  76995-602
 
 :   Ambeed
 :  A118216-100G
Vanillin 99%
  76995-602
 :  Ambeed
 :  121-33-5
Vanillin 99%

Glass bottle, 100 g

 :  121-33-5
 
$26.00
   
Each (100g)
 
   
 

 : for Research Use Only

 :  4-Hydroxy-3-methoxybenzaldehyde
  •  :121-33-5
  •  :C₈H₈O₃
  •  :MFCD00006942
  •  :152.15 g/mol
 

 

  • Pack type:
    Glass bottle
  • Size:
    100 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.