Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Vanillin 99%

Vanillin 99%

Glass bottle, 1000 g
Catalog # 76995-600
Irritant
Supplier:   Ambeed
Supplier #:  A118216-1000G
Vanillin 99%
Catalog # 76995-600
Supplier:  Ambeed
CAS Number:  121-33-5
Vanillin 99%

Glass bottle, 1000 g

CAS Number:  121-33-5
Price
$46.00
Log in for availability
Each (1,000g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

 : for Research Use Only

Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1000 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.