Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SDS-SAMPLE LAEMMLI 6X REDUCE 50ML
Catalog # MSPP-BP111R50M
Supplier:
BOSTON BIOPRODUCTS MS
Expand image
BUFFER SDS-SAMPLE LAEMMLI 6X REDUCE 50ML
Catalog # MSPP-BP111R50M
Supplier:
BOSTON BIOPRODUCTS MS
Supplier Number:
BP-111R-50ML