Accessories for Level Control Signal Boxes
Power supply-USA 110 V for signal box T1 and T5, 24 Volt, cable length: 1900 mm
Catalog # 76948-984
Supplier: SCAT AMERICAS, INC.
Supplier #: 610704
Jump to:
Product Details & Documents
Useful accessories, extensions, and alternative products for the SCAT level control signal boxes.
- Cable for relay contacts
- Disc sensors
- Dual lock
- Signal cables
- Power supplies
- Velcro strips
Ensure SCAT level control signal boxes are perfectly equipped.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Description:Power supply-USA 110 V for signal box T1 and T5, 24 Volt, cable length: 1900 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



