Accessories for Level Control Signal Boxes
Air valve for SafetyCaps V 2.0, UNF ¹/₄" 28G, service life: 6 months
Catalog # 76948-972
Marketsource
Supplier: SCAT AMERICAS, INC.
Supplier #: 397100
Accessories for Level Control Signal Boxes
Catalog # 76948-972
Supplier: SCAT AMERICAS, INC.
Marketsource
Jump to:
Product Details & Documents
Useful accessories, extensions, and alternative products for the SCAT level control signal boxes.
- Cable for relay contacts
- Disc sensors
- Dual lock
- Signal cables
- Power supplies
- Velcro strips
Ensure SCAT level control signal boxes are perfectly equipped.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Description:Air valve for SafetyCaps V 2.0, UNF ¹/₄" 28G, service life: 6 months
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



