Orders LinkContactUsLinkComponent
Control Slides, Normal Human Tissue, Prostate
Control slides, normal human tissue, prostate
  76948-206
 :   Mopec
 :  SC942-100
Control Slides, Normal Human Tissue, Prostate
  76948-206
 :  Mopec
Control Slides, Normal Human Tissue, Prostate
Control Slides, Normal Human Tissue, Prostate
Control slides, normal human tissue, prostate
$1,300.49
Case of 100
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
 : Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.
  • Description:
    Control slides, normal human tissue, prostate